Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HBsAg Antibody (Hs33), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10073138AF350
Description
HBsAg Monoclonal antibody specifically detects HBsAg in Virus samples. It is validated for ELISASpecifications
HBsAg | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Virus | |
Purified |
Sandwich ELISA | |
Hs33 | |
Sandwich ELISA | |
HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with recombinant HBsAg of ayw subtype. | |
0.2 mL | |
Cell Biology, Virology Bacteria and Parasites | |
944569 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction