Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBsAg Antibody, Novus Biologicals™
SDP

Catalog No. NB017503 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017503 0.1 mL
NB10062652 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017503 Supplier Novus Biologicals Supplier No. NB100626520.1ML
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HBsAg Polyclonal specifically detects HBsAg in Human, Mouse, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen HBsAg
Applications Immunohistochemistry (Frozen), ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Polyclonal
Concentration 4.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen
Formulation PBS with 0.1% Sodium Azide
Gene Alias HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S
Gene Symbols S
Host Species Rabbit
Immunogen Hepatitis B subtypes ad and ay.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 944569
Test Specificity This product is specific for Hepatitis B surface antigen, recognizing subtypes ad and ay. Hepatitis B virus is a major causative agent of acute and chronic liver disease in humans. Hepatitis B surface antigen (HBsAg) is a protein component of the viral envelope, which is expressed predominantly in the cytoplasm of infected hepatocytes.
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.