Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBTU 98.0+%, TCI America™
SDP

Catalog No. B165725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B165725G 25 g
B16575G 5 g
B1657100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B165725G Supplier TCI America Supplier No. B165725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 94790-37-1
Molecular Formula C11H16F6N5OP
Molecular Weight (g/mol) 379.247
MDL Number MFCD00075445
InChI Key UQYZFNUUOSSNKT-UHFFFAOYSA-N
Synonym hbtu, o-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, 2-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate, hbtu-reagent, o-benzotriazol-1-yl-tetramethyluronium hexafluorophosphate, hbtu o-benzotriazole-n,n,n',n'-tetramethyl-uronium-hexafluoro-phosphate, n,n,n',n'-tetramethyl-o-1h-benzotriazol-1-yl uronium hexafluorophosphate, o-1h-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, o-benzotriazole-n,n,n',n'-tetramethyluronium hexafluorophosphate
PubChem CID 2733084
IUPAC Name [benzotriazol-1-yloxy(dimethylamino)methylidene]-dimethylazanium;hexafluorophosphate
SMILES CN(C)C(=[N+](C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F

Specifications

Color White
Quantity 25 g
Formula Weight 379.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material HBTU [Coupling Reagent for Peptide]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.