Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ HBV X (154 amino acids) Recombinant Protein

Catalog No. 89947134 Shop All Abnova Corporation Products
Click to view available options
Quantity:
50 μg

Used for SDS-PAGE

Sequence: MAARVCCQLDPARDVLCLRPVGAESRGRPVSGPFGTLPSPSSSAVPADHGAHLSLRGLPVCAFSSAGPCALRFTSARRMETTVNAHQVLPKVLHKRTLGLSAMSTTDLEAYFKDCLFKDWEELGEEIRLKVFVLGGCRHKLVCSPAPCNFFTSA

Specifications

For Use With (Application) SDS-PAGE
Formulation Liquid
Molecular Weight (g/mol) 17kDa
Name HBV (X) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 50 μg
Immunogen MAARVCCQLDPARDVLCLRPVGAESRGRPVSGPFGTLPSPSSSAVPADHGAHLSLRGLPVCAFSSAGPCALRFTSARRMETTVNAHQVLPKVLHKRTLGLSAMSTTDLEAYFKDCLFKDWEELGEEIRLKVFVLGGCRHKLVCSPAPCNFFTSA
Storage Requirements Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name HBV
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.