Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HBXIP Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | HBXIP |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
HBXIP Polyclonal specifically detects HBXIP in Human, Mouse samples. It is validated for Western Blot.Specifications
HBXIP | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
HBV X-interacting protein, HBX-interacting protein, hepatitis B virus x interacting protein, hepatitis B virus x-interacting protein (9.6kD), MGC71071, XIPhepatitis B virus X-interacting protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-173 of human HBXIP (NP_006393.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Stem Cell Markers | |
PBS with 50% glycerol, pH7.3. | |
10542 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title