Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HBZ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584872
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584872 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584872 Supplier Invitrogen™ Supplier No. PA584872
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TIIVSMWAKI STQADTIGTE TLERLFLSHP QTKTY Highest antigen sequence indentity to the following orthologs: Mouse - 71%, Rat - 69%.

This gene encodes a member of a subfamily of lipid trafficking proteins that are characterized by a C-terminal steroidogenic acute regulatory domain and an N-terminal metastatic lymph node 64 domain. The encoded protein localizes to the membranes of late endosomes and may be involved in exporting cholesterol. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HBZ
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Hbz
Gene Accession No. P02008
Gene Alias AI450015; alpha-like embryonic globin chain x; Hba-x; HBAZ; HBZ; HBZ1; HBZ2; HBZ-T1; hemaglobin zeta; hemoglobin subunit zeta; hemoglobin X, alpha-like embryonic chain in Hba complex; Hemoglobin zeta; hemoglobin zeta chain; hemoglobin, zeta; RA_M008_JSM295ECF; RGD1307486; zeta globin; zeta-globin
Gene Symbols Hbz
Host Species Rabbit
Immunogen Recombinant protein fragment of Human HBZ (Product #RP-108588)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3050
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.