Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HBZ Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593287
Description
Immunogen sequence: MSLTKTERTI IVSMWAKIST QADTIGTETL ERLFLSHPQT KTYFPHFDLH PGSAQLRAHG SKVVAAVGDA VKSIDDIGGA LSKLSELHAY ILRVDPVNFK LLSHCLLVTL AARFPADFTA EAHAAWDKFL SVVSSVLTEK YR; Positive Samples: K-562.
This gene encodes a member of a subfamily of lipid trafficking proteins that are characterized by a C-terminal steroidogenic acute regulatory domain and an N-terminal metastatic lymph node 64 domain. The encoded protein localizes to the membranes of late endosomes and may be involved in exporting cholesterol. Alternative splicing results in multiple transcript variants.
Specifications
| HBZ | |
| Polyclonal | |
| Unconjugated | |
| Hbz | |
| AI450015; alpha-like embryonic globin chain x; Hba-x; HBAZ; HBZ; HBZ1; HBZ2; HBZ-T1; hemaglobin zeta; hemoglobin subunit zeta; hemoglobin X, alpha-like embryonic chain in Hba complex; Hemoglobin zeta; hemoglobin zeta chain; hemoglobin, zeta; RA_M008_JSM295ECF; RGD1307486; zeta globin; zeta-globin | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3050 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.67 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02008 | |
| Hbz | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-142 of human HBZ (NP_0053231). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction