Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HBZ Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139861
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: AHGSKVVAAV GDAVKSIDDI GGALSKLSEL HAYILR.
This gene encodes a member of a subfamily of lipid trafficking proteins that are characterized by a C-terminal steroidogenic acute regulatory domain and an N-terminal metastatic lymph node 64 domain. The encoded protein localizes to the membranes of late endosomes and may be involved in exporting cholesterol. Alternative splicing results in multiple transcript variants.
Specifications
HBZ | |
Polyclonal | |
Unconjugated | |
Hbz | |
AI450015; alpha-like embryonic globin chain x; Hba-x; HBAZ; HBZ; HBZ1; HBZ2; HBZ-T1; hemaglobin zeta; hemoglobin subunit zeta; hemoglobin X, alpha-like embryonic chain in Hba complex; Hemoglobin zeta; hemoglobin zeta chain; hemoglobin, zeta; RA_M008_JSM295ECF; RGD1307486; zeta globin; zeta-globin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3050 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P02008 | |
Hbz | |
Recombinant protein corresponding to Human HBZ. Recombinant protein control fragment (Product #RP-109035). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction