Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HBZ Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | HBZ |
|---|---|
| Concentration | 0.19 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2644
![]()
|
Proteintech Group Inc
199951AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody is specific to HBZ.
Zeta-globin is an alpha-like hemoglobin. The zeta-globin polypeptide is synthesized in the yolk sac of the early embryo, while alpha-globin is produced throughout fetal and adult like. The zeta-globin gene is a member of the human alpha-globin gene cluster that includes five functional genes and two pseudogenes. The order of genes is: 5′ - zeta - pseudozeta - mu - pseudoalpha-1 - alpha-2 -alpha-1 - theta1 - 3′. Zeta-globin (HBZ ) is an alpha-like hemoglobin. The zeta-globin polypeptide is synthesized in the yolk sac of the early embryo, while alpha-globin is produced throughout fetal and adult like. The zeta-globin gene is a member of the human alpha-globin gene cluster that involves 4 functional genes and 3 nonfunctional pseudogenes. The order of genes is: 5′-zeta -- pseudozeta -- pseudoalpha2 -- pseudoalpha1 -- alpha2 -- alph1 -- theta1-3′.Specifications
| HBZ | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P02008 | |
| 3050 | |
| Peptide | |
| Primary | |
| -20°C | |
| HBZ |
| 0.19 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| HBAP2, HBAZ, HBM, HBZ, HBZ2, Hemoglobin mu chain, Hemoglobin subunit mu, Hemoglobin subunit zeta, Hemoglobin zeta chain, hemoglobin, mu, hemoglobin, zeta, Mu globin, Zeta globin | |
| HBZ | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title