Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCA66 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572598
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2598 10 μL
50-157-2597 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2598 Supplier Bethyl Laboratories, Inc Supplier No. IHC00719T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Monkey, Gorilla, Northern white-cheeked gibbon, Crab-eating macaque and White-tufted-e

UTP6 involved in nucleolar processing of pre-18S ribosomal RNA.

Specifications

Antigen HCA66
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene UTP6
Gene Accession No. Q9NYH9
Gene Alias C17orf40, HCA66, U3 small nucleolar RNA-associated protein 6 homolog, hepatocellular carcinoma associated antigen 66, hepatocellular carcinoma-associated antigen 66, multiple hat domains protein
Gene Symbols UTP6
Host Species Rabbit
Immunogen Between 565 and 597
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55813
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.