Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCAP G Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983753
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3753 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3753 Supplier Bioss Supplier No. BS15422R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a subunit of the condensin complex, which is responsible for the condensation and stabilization of chromosomes during mitosis and meiosis. Phosphorylation of the encoded protein activates the condensin complex. There are pseudogenes for this gene on chromosomes 8 and 15. Alternative splicing results in multiple transcript variants.

Specifications

Antigen HCAP G
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NCAPG
Gene Accession No. Q9BPX3
Gene Alias 5730507H05Rik; CAPG; CHCG; chromosome condensation protein G; Chromosome-associated protein G; Condensin complex subunit 3; Condensin subunit CAP-G; FLJ12450; Hcapg; hCAP-G; Melanoma antigen NY-MEL-3; MFT.M05.13; MGC126525; NCAPG; Non-SMC condensin I complex subunit G; non-SMC condensin I complex, subunit G; NYMEL3; NY-MEL-3; on-SMC condensin I complex, subunit G; XCAP-G homolog; YCG1
Gene Symbols NCAPG
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HCAP G.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64151
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.