Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HCAR2/HCAR3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599937
Description
Antibody detects endogenous levels of total GPR109A/B.
Acts as a high affinity receptor for both nicotinic acid (also known as niacin) and (D)-beta-hydroxybutyrate and mediates increased adiponectin secretion and decreased lipolysis through G(i)-protein-mediated inhibition of adenylyl cyclase. This pharmacological effect requires nicotinic acid doses that are much higher than those provided by a normal diet. Mediates nicotinic acid-induced apoptosis in mature neutrophils. Receptor activation by nicotinic acid results in reduced cAMP levels which may affect activity of cAMP-dependent protein kinase A and phosphorylation of target proteins, leading to neutrophil apoptosis. The rank order of potency for the displacement of nicotinic acid binding is 5-methyl pyrazole-3-carboxylic acid = pyridine-3-acetic acid > acifran > 5-methyl nicotinic acid = acipimox >> nicotinuric acid = nicotinamide.
Specifications
HCAR2/HCAR3 | |
Polyclonal | |
Unconjugated | |
HCAR3 | |
G protein-coupled receptor 109A; G protein-coupled receptor 109B; G protein-coupled receptor HM74a; GPR109A; GPR109B; G-protein coupled receptor 109A; G-protein coupled receptor 109B; G-protein coupled receptor HM74; G-protein coupled receptor HM74A; G-protein coupled receptor HM74B; GTP-binding protein; HCA2; HCA3; HCAR2; HCAR3; HM74; HM74a; HM74B; hydroxycarboxylic acid receptor 2; hydroxy-carboxylic acid receptor 2; Hydroxycarboxylic acid receptor 3; hydroxy-carboxylic acid receptor 3; niacin receptor 1; Niacin receptor 2; NIACR1; NIACR2; Nicotinic acid receptor; nicotinic acid receptor 2; PUMAG; Puma-g; putative chemokine receptor | |
Rabbit | |
Affinity chromatography | |
RUO | |
338442, 8843 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P49019, Q8TDS4 | |
HCAR2, HCAR3 | |
A synthesized peptide derived from human HCAR2(Accession Q8TDS4), corresponding to amino acid residues F304-S354. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction