Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HCC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590681
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590681 Supplier Invitrogen™ Supplier No. PA590681
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATQCFQLSNM FNPQTEEEVG WDTEIKDDVI EECNKHGGVI HIYVDKNSAQ GNVYVKCPSI AAAIAAVNAL HGRWFAGKMI TAAYVPLPTY HNLFPDSMTA TQLLVPSRR; Positive Samples: BxPC-3, U-251MG, 293T, LO2, Mouse brain, Mouse heart; Cellular Location: Nucleus speckle.

The protein encoded by this gene is an RNA binding protein and possible splicing factor. The encoded protein is found in the nucleus, where it colocalizes with core spliceosomal proteins. Studies of a mouse protein with high sequence similarity to this protein suggest that this protein may act as a transcriptional coactivator for JUN/AP-1 and estrogen receptors. Multiple transcript variants encoding different isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCC1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RBM39
Gene Accession No. Q14498, Q8VH51
Gene Alias 1500012C14Rik; 2310040E03Rik; B330012G18Rik; C79248; CAPER; CAPER alpha; CAPERalpha; Coactivator of activating protein 1 and estrogen receptors; coactivator of activating protein-1 and estrogen receptors; coactivator of AP-1 and ERs; FSAP59; functional spliceosome-associated protein 59; HCC1; Hepatocellular carcinoma protein 1; R75070; RBM39; RNA binding motif protein 39; RNA-binding motif protein 39; RNA-binding protein 39; RNA-binding region (RNP1, RRM) containing 2; RNA-binding region-containing 2; RNA-binding region-containing protein 2; Rnp1; Rnpc2; Rrm; Splicing factor HCC1; transcription coactivator CAPER
Gene Symbols RBM39
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170791, 362251, 9584
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.