Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCCS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106551
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106551 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106551 Supplier Thermo Scientific Supplier No. PA5106551
Add to Cart
Add to Cart

HCCS Polyclonal Antibody

HCCS Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen HCCS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HCCS
Gene Accession No. P53701, P53702
Gene Alias CCHL; cytochrome c heme-lyase; cytochrome c-type heme lyase; HCCS; holocytochrome c synthase; holocytochrome c synthetase; holocytochrome c-type synthase; LSDMCA1; MCOPS7; microphthalamia with linear skin defects; MLS; RGD1563855
Gene Symbols HCCS
Host Species Rabbit
Immunogen A synthesized peptide derived from human HCCS(Accession P53701), corresponding to amino acid residues L84-G134.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15159, 3052, 317444
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.