Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCE Polyclonal Antibody
HCE Polyclonal Antibody
Supplier: Thermo Scientific PA598371
Description
HCE Polyclonal Antibody for Western Blot, IHC (P), ELISA

Specifications
HCE | |
Polyclonal | |
Unconjugated | |
RNGTT | |
AU020997; CAP1A; GTase; GTP--RNA guanylyltransferase; hCAP; HCAP1; HCE; HCE1; MCE1; mRNA 5'-triphosphatase; mRNA guanylyltransferase; mRNA-capping enzyme; Polynucleotide 5'-triphosphatase; RNA guanylyltransferase and 5'-phosphatase; Rngtt; RP3-488C13.1; TPase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8732 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60942 | |
RNGTT | |
Recombinant Human mRNA-capping enzyme protein (1-260AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction