Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HCE Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521684

Catalog No. PIPA521684


Only null left
Add to Cart

Description

Description

Recommended positive controls: 293T, A431, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (95%), Rat (96%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Bifunctional mRNA-capping enzyme exhibiting RNA 5'-triphosphatase activity in the N-terminal part and mRNA guanylyltransferase activity in the C-terminal part. Catalyzes the first two steps of cap formation: by removing the gamma-phosphate from the 5'-triphosphate end of nascent mRNA to yield a diphosphate end, and by transferring the gmp moiety of GTP to the 5'-diphosphate terminus.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HCE
Polyclonal
Unconjugated
RNGTT
AU020997; CAP1A; GTase; GTP--RNA guanylyltransferase; hCAP; HCAP1; HCE; HCE1; MCE1; mRNA 5'-triphosphatase; mRNA guanylyltransferase; mRNA-capping enzyme; Polynucleotide 5'-triphosphatase; RNA guanylyltransferase and 5'-phosphatase; Rngtt; RP3-488C13.1; TPase
Rabbit
Antigen affinity chromatography
RUO
8732
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.54 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O60942
RNGTT
Recombinant protein encompassing a sequence within the center region of human HCE. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.