Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCE Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983755
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3755 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3755 Supplier Bioss Supplier No. BS15425R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Bifunctional mRNA-capping enzyme exhibiting RNA 5′-triphosphatase activity in the N-terminal part and mRNA guanylyltransferase activity in the C-terminal part. Catalyzes the first two steps of cap formation: by removing the gamma-phosphate from the 5′-triphosphate end of nascent mRNA to yield a diphosphate end, and by transferring the gmp moiety of GTP to the 5′-diphosphate terminus.

Specifications

Antigen HCE
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RNGTT
Gene Accession No. O60942
Gene Alias AU020997; Cap1a; GTase; GTP--RNA guanylyltransferase; hCAP; HCAP1; HCE; HCE1; MCE1; mRNA 5′-triphosphatase; mRNA guanylyltransferase; mRNA-capping enzyme; Polynucleotide 5′-triphosphatase; RNA guanylyltransferase and 5′-phosphatase; RNGTT; RP3-488C13.1; TPase
Gene Symbols RNGTT
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HCE.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8732
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.