Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HCFC1R1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563761
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563761 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563761 Supplier Invitrogen™ Supplier No. PA563761
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QPLQRGPQGG AQRLPRAALG VTWGLDASSP LRGAVPMSTK RRLEEEQEPL RKQFLSEEN Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 46%.

Regulates HCFC1 activity by modulating its subcellular localization. Overexpression of HCFC1R1 leads to accumulation of HCFC1 in the cytoplasm. HCFC1R1-mediated export may provide the pool of cytoplasmic HCFC1 required for import of virion-derived VP16 into the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCFC1R1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HCFC1R1
Gene Accession No. Q9NWW0
Gene Alias HCF-1 beta-propeller-interacting protein; HCFC1R1; Host cell factor C1 regulator 1; host cell factor C1 regulator 1 (XPO1 dependent); HPIP
Gene Symbols HCFC1R1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HCFC1R1. Recombinant protein control fragment (Product #RP-101727).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54985
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.