Learn More
HCFC2 Mouse anti-Human, Clone: OTI4E11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505237

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes one of two proteins which interact with VP16, a herpes simplex virus protein that initiates virus infection. Both the encoded protein and the original Herpes host cell factor interact with VP16 through a beta-propeller domain. The original Herpes host cell factor, however, is effective at initiating viral infection while the encoded protein is not. Transcripts of varying length due to alternative polyadenylation signals have been described.Specifications
HCFC2 | |
Monoclonal | |
Unconjugated | |
HCFC2 | |
HCF-2, HCF2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
29915 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4E11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y5Z7 | |
HCFC2 | |
Full length human recombit protein of human HCFC2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.