Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HCFC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552267
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552267 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552267 Supplier Invitrogen™ Supplier No. PA552267
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APSQVQLIKA TTNSFHVKWD EVSTVEGYLL QLSTDLPYQA ASSDSSAAPN MQGVRMDPHR QGSNNIVPNS INDTINSTKT EQPATKETSM KNKPDFKALT DSNAILYPSL ASNASNHNSH VVDMLRKNEG PHTSANVGV Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 60%.

This gene encodes one of two proteins which interact with VP16, a herpes simplex virus protein that initiates virus infection. Both the encoded protein and the original Herpes host cell factor interact with VP16 through a beta-propeller domain. The original Herpes host cell factor, however, is effective at initiating viral infection while the encoded protein is not. Transcripts of varying length due to alternative polyadenylation signals have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCFC2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HCFC2
Gene Accession No. Q9Y5Z7
Gene Alias 1700129L13Rik; AU019838; C2 factor; HCF2; HCF-2; HCFC2; Host cell factor 2; host cell factor C2
Gene Symbols HCFC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HCFC2. Recombinant protein control fragment (Product #RP-89327).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29915
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.