Learn More
Invitrogen™ hCG_1657980 Polyclonal Antibody

Description
hCG_1657980 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | hCG_1657980 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Host Species | Rabbit |
| Immunogen | KLH conjugated synthetic peptide between 94-122 amino acids from the Central region of human hCG_1657980. |
| Purification Method | Antigen Affinity Chromatography, Protein A |
| Quantity | 400 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.