Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hCG Antibody, DyLight 550, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP311024R
Description
hCG Polyclonal antibody specifically detects hCG in Human samples. It is validated for Western Blot, ELISASpecifications
hCG | |
Polyclonal | |
Western Blot, ELISA | |
Goat | |
Affinity purified | |
RUO | |
Human | |
Purified |
Western Blot, ELISA | |
DyLight 550 | |
50mM Sodium Borate | |
Human TSH (intact) | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction