Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hCG beta-1 Monoclonal Antibody (INN-hCG-2)
GREENER_CHOICE

Catalog No. PIMA516740
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516740 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516740 Supplier Invitrogen™ Supplier No. MA516740
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti hCG antibody, clone INN-hCG-2 recognizes the beta 1 epitope on human chorionic gonadotrophin (hCG).Chorionic gonadotrophin is a member of the glycoprotein hormone family that is synthesized by the placenta throughout pregnancy.

HCG is a glycoprotein, which is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the alpha and Beta subunits. The alpha subunit is identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH).
TRUSTED_SUSTAINABILITY

Specifications

Antigen hCG beta-1
Applications ELISA, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA)
Classification Monoclonal
Clone INN-hCG-2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene CGB1
Gene Accession No. A6NKQ9
Gene Alias CGB1; CGB3; CGB5; CGB7; CGB8; CG-beta; Choriogonadotropin subunit beta variant 1; chorionic gonadotropin beta subunit 1; chorionic gonadotropin human; chorionic gonadotropin, beta polypeptide 1; hCGB; human chorionic gonadotropin
Gene Symbols CGB1
Host Species Mouse
Immunogen hCG.
Purification Method Protein A
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114335
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.