Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hCG beta-2 Monoclonal Antibody (INN-hCG-22)
GREENER_CHOICE

Catalog No. PIMA516741
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516741 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516741 Supplier Invitrogen™ Supplier No. MA516741
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Human chorionic gonadotrophin antibody, clone INN-hCG-22 recognizes the beta subunit of human choriogonadotrophin (hCG), also known as chorionic gonadotrophin.

The beta subunit of chorionic gonadotropin (CGB) is encoded by six highly homologous and structurally similar genes that are arranged in tandem and inverted pairs on chromosome 19q13.3, and contiguous with the luteinizing hormone beta (LHB) subunit gene. The CGB genes are primarily distinguished by differences in the 5' untranscribed region. This gene was originally thought to be one of the two pseudogenes (CGB1 and CGB2) of CGB subunit, however, detection of CGB1 and CGB2 transcripts in vivo, and their presence on the polysomes, suggested that these transcripts are translated. To date, a protein product corresponding to CGB2 has not been isolated. The deduced sequence of the hypothetical protein of 132 aa does not share any similarity with that of functional CGB subunits (PMID:8954017). However, a 163 aa protein, translated from a different frame, is about the same size, and shares 98% identity with other CGB subunits. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen hCG beta-2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone INN-hCG-22
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene CGB2
Gene Accession No. Q6NT52
Gene Alias CGB2; CGB3; CGB5; CGB7; CGB8; CG-beta; Choriogonadotropin subunit beta variant 2; chorionic gonadotropin beta subunit 2; chorionic gonadotropin human; chorionic gonadotropin, beta polypeptide 2; hCGB; human chorionic gonadotropin; product of CGB2
Gene Symbols CGB2
Host Species Mouse
Immunogen hCG.
Purification Method Protein A
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114336
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.