Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ hCG beta-4 Monoclonal Antibody (INN-hCG-24)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA125110
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Human chorionic gonadotropin (hCG) is a glycoprotein hormone produced by trophoblastic cells of the placenta beginning 10 to 12 days after conception. Maintenance of the fetus in the first trimester of pregnancy requires the production of hCG, which binds to the corpus luteum of the ovary which is stimulated to produce progesterone which in turn maintains the secretory endometrium.
Specifications
hCG beta-4 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P0DN86 | |
CGB3 | |
Human Chorionic Gonadotropin. | |
250 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Radioimmune Assays (RIA) | |
INN-hCG-24 | |
Unconjugated | |
CGB3 | |
CGB; CGB3; CGB5; CGB7; CGB8; CG-beta; Choriogonadotropin subunit beta; choriogonadotropin subunit beta 3; chorionic gonadotrophin chain beta; chorionic gonadotropin beta 3 subunit; chorionic gonadotropin beta chain; chorionic gonadotropin beta subunit; chorionic gonadotropin beta subunit 3; chorionic gonadotropin chain beta; chorionic gonadotropin human; chorionic gonadotropin, beta polypeptide; hCGB; hCG-beta; human chorionic gonadotropin | |
Mouse | |
Protein G | |
RUO | |
1082 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction