Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hCG beta 7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30357520UL
Description
hCG beta 7 Polyclonal specifically detects hCG beta 7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
hCG beta 7 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
Unconjugated | |
CGB3, CG-beta, CG-beta-a, Chorionic gonadotrophin chain beta, chorionic gonadotropin beta 7 subunit, chorionic gonadotropin, beta polypeptide 7, FLJ35403, FLJ43118 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 21-165 of human hCG beta 7 (NP_149133.1). | |
20 μL | |
Apoptosis, Cancer | |
94027 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction