Learn More
Invitrogen™ hCG beta Monoclonal Antibody (2092)

Description
The antibody does stain HCG containing human cells.
Specifications
Specifications
| Antigen | hCG beta |
| Applications | ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 2092 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Tissue culture supernatant with 10mM sodium azide |
| Gene | CGB3 |
| Gene Accession No. | P0DN86 |
| Gene Alias | CGB; CGB3; CGB5; CGB7; CGB8; CG-beta; Choriogonadotropin subunit beta; choriogonadotropin subunit beta 3; chorionic gonadotrophin chain beta; chorionic gonadotropin beta 3 subunit; chorionic gonadotropin beta chain; chorionic gonadotropin beta subunit; chorionic gonadotropin beta subunit 3; chorionic gonadotropin chain beta; chorionic gonadotropin human; chorionic gonadotropin, beta polypeptide; FSH-alpha; hCGB; human chorionic gonadotropin; LSH-alpha; TSH-alpha |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.