Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hCG beta Mouse anti-Human, Clone: 2D6B8, Proteintech
SDP

Catalog No. 501736421 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6421 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6421 Supplier Proteintech Group Inc Supplier No. 603341IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Human chorionic gonadotropin (hCG) is a placental hormone that stimulates secretion of the pregcy-sustaining steroid progesterone. hCG is a member of a family of glycoprotein hormones including pituitary hormones lutropin (luteinizing hormone;LH), follitropin (follicle-stimulating hormone; FSH) and thyrotropin (thyroid-stimulating hormone; TSH). Members of this family are all composed of disulfide-rich heterodimers with two dissimilar subunits namely alpha- and beta-subunits. The alpha subunit is common to all the members of the family within a given species. The beta subunits are distinct in each of the horomones and confer the receptor and biological specificity.

Specifications

Antigen hCG beta
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2D6B8
Concentration 0.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CGB3
Gene Accession No. P0DN86
Gene Alias CG beta, CGB, CGB,hCG, CGB3, CGB5, CGB7, CGB8, hCG, hCGB
Gene Symbols CGB3
Host Species Mouse
Immunogen hCG beta Fusion Protein Ag2191
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1082
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.