Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hCG beta Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501727312 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7312 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7312 Supplier Proteintech Group Inc Supplier No. 116151AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Human chorionic gonadotropin (hCG) is a placental hormone that stimulates secretion of the pregcy-sustaining steroid progesterone. hCG is a member of a family of glycoprotein hormones includes pituitary hormones lutropin (luteinizing hormone;LH), follitropin (follicle-stimulating hormone; FSH) and thyrotropin (thyroid-stimulating hormone; TSH). Members of this family are all composed of disulfide-rich heterodimers with two dissimilar subunits namely alpha- and beta-subunits. Alpha subunit, which is common to all the members of the family within a given species, with distinct but homologous beta subunits which confer hormone specificities.

Specifications

Antigen hCG beta
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CGB3
Gene Accession No. P0DN86
Gene Alias CG beta, CGB, CGB3, CGB5, CGB7, CGB8, hCG, hCGB
Gene Symbols CGB3
Host Species Rabbit
Immunogen hCG beta Fusion Protein Ag2191
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1082
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.