Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hCG Holo c3 Monoclonal Antibody (INN-hCG-45)
GREENER_CHOICE

Catalog No. PIMA125112
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA125112 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA125112 Supplier Invitrogen™ Supplier No. MA125112
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-25112 detects hCG Holo c3 from human samples.

Human chorionic gonadotropin (hCG) is a glycoprotein hormone produced by trophoblastic cells of the placenta beginning 10 to 12 days after conception. Maintenance of the fetus in the first trimester of pregnancy requires the production of hCG, which binds to the corpus luteum of the ovary which is stimulated to produce progesterone which in turn maintains the secretory endometrium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hCG Holo c3
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Clone INN-hCG-45
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Alias CGB; CGB3; CGB5; CGB7; CGB8; CG-beta; choriogonadotropin subunit beta; Choriogonadotropin subunit beta 3; chorionic gonadotrophin chain beta; chorionic gonadotropin beta 3 subunit; chorionic gonadotropin beta chain; chorionic gonadotropin beta subunit; chorionic gonadotropin beta subunit 3; chorionic gonadotropin chain beta; chorionic gonadotropin human; chorionic gonadotropin, beta polypeptide; hCGB; human chorionic gonadotropin
Host Species Mouse
Immunogen Human Chorionic Gonadotropin.
Purification Method Protein A
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.