Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hCG Monoclonal Antibody (3015)
GREENER_CHOICE

Catalog No. PIMA514700
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514700 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514700 Supplier Invitrogen™ Supplier No. MA514700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The observed molecular weight of alpha-hCG is approximately 20-22 kD. The molecular weight is higher than the predicted molecular weight due to post translational modification. Product MA514700 is a smaller package size of MIH9805 (formerly sold as a Seradyn product).

Human chorionic gonadotropin (hCG) is a placental hormone that stimulates secretion of the pregnancy-sustaining steroid progesterone. hCG is a member of a family of glycoprotein hormones including pituitary hormones lutropin (luteinizing hormone;LH), follitropin (follicle-stimulating hormone; FSH) and thyrotropin (thyroid-stimulating hormone; TSH). Members of this family are all composed of disulfide-rich heterodimers with two dissimilar subunits namely alpha- and beta-subunits. The alpha subunit is common to all the members of the family within a given species. The beta subunits are distinct in each of the horomones and confer the receptor and biological specificity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hCG
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone 3015
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Gene CGB3
Gene Accession No. P01215, P0DN86
Gene Alias anterior pituitary glycoprotein hormones common subunit alpha; CGA; CG-alpha; CGB; CGB3; CGB5; CGB7; CGB8; CG-beta; Choriogonadotropin alpha chain; choriogonadotropin subunit beta; Choriogonadotropin subunit beta 3; chorionic gonadotrophin chain beta; chorionic gonadotrophin subunit alpha; chorionic gonadotropin beta 3 subunit; chorionic gonadotropin beta 5 subunit; chorionic gonadotropin beta 8 subunit; chorionic gonadotropin beta chain; chorionic gonadotropin beta subunit; chorionic gonadotropin beta subunit 3; chorionic gonadotropin beta subunit 5; chorionic gonadotropin beta subunit 8; chorionic gonadotropin chain beta; chorionic gonadotropin human; chorionic gonadotropin, alpha polypeptide; chorionic gonadotropin, beta polypeptide; chorionic gonadotropin, beta polypeptide 5; chorionic gonadotropin, beta polypeptide 8; follicle-stimulating hormone alpha chain; follicle-stimulating hormone alpha subunit; follitropin alpha chain; FSHA; FSH-alpha; Glycoprotein hormones alpha chain; glycoprotein hormones, alpha polypeptide; GPHa; GPHA1; HCG; hCGB; human chorionic gonadotropin; LHA; LSH-alpha; luteinizing hormone alpha chain; Lutropin alpha chain; thyroid-stimulating hormone alpha chain; Thyrotropin alpha chain; TSHA; TSH-alpha
Gene Symbols CGA, CGB3, CGB5, CGB8
Host Species Mouse
Immunogen Human Chorionic Gonadotropin.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1081, 1082, 93659, 94115
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 6 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.