Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hCG receptor Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980613
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0613 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0613 Supplier Bioss Supplier No. BS0984R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LHCGR encodes the receptor for both luteinizing hormone and choriogonadotropin. It is family 1 of G-protein coupled receptors, FSH/LSH/TSH superfamily, and its basic function is acting as luteinizing hormone/choriogonadotropin receptor (gonadotropin receptor). Regulating the development and function of Leyding cells in males. It also plays a role in male secondary sexual characterdevelopment. Mutations in this gonadotropinreceptor results in disorders of male secondary sexual characteristic development, such as familial male precocious puperty and male pseudohermaphtoditism.

Specifications

Antigen hCG receptor
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LHCGR
Gene Accession No. P22888
Gene Alias Gpcr19-rs1; HCG receptor; HHG; hypergonadotropic hypogonadism; LCGR; LGR2; LH receptor; LH/CGR; LH/CG-R; LHCGR; Lhr; LH-R; LHRHR; LSHR; LSH-R; Luteinizing hormone receptor; luteinizing hormone/choriogonadotropin receptor; Lutropin receptor; lutropin/choriogonadotropin receptor; Lutropin-choriogonadotropic hormone receptor; ULG5
Gene Symbols LHCGR
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse LHCGR.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3973
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.