Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCG3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP243615
Description
Specifically detects HCG3 in Human samples, and it is validated for Western Blotting
HCG3 Polyclonal specifically detects HCG3 in Human samples. It is validated for Western Blot.Specifications
HCG3 | |
Polyclonal | |
Western Blot 1:500-1:3000 | |
DnaJ (Hsp40) homolog, subfamily B, member 3, dnaJ homolog subfamily B member 3, HCG3MGC26879 | |
Rabbit | |
17 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
DNAJB3 | |
Recombinant protein encompassing a sequence within the center region of human DNAJB3. The exact sequence is proprietary. | |
Immunogen affinity purified | |
RUO | |
414061 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction