Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HCG9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570679
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570679 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570679 Supplier Invitrogen™ Supplier No. PA570679
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

This gene lies within the MHC class I region on chromosome 6p21.3. This gene is believed to be non-coding, but its function has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCG9
Applications Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HCGIX
Gene Accession No. 0
Gene Alias HCG9; HCGIX; HCGIX4; HLA complex group 9 (non-protein coding)
Gene Symbols HCGIX
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human HCG9.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10255
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.