Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human HCK, GST Tag Recombinant Protein

Catalog No. PV6128
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV6128 10 μg
PR9171A 100 μg
PV6129 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PV6128 Supplier Invitrogen™ Supplier No. PV6128
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Hematopoietic cell kinase (HCK) is a SRC-related kinase expressed in lymphoid and hematopoietic cells and may play a role in some forms of leukemia. (HCK Alias: DKFZp686D0638). HCK is a protein-tyrosine kinase that is predominantly expressed in hemopoietic cell types. The encoded protein may help couple the Fc receptor to the activation of the respiratory burst. In addition, it may play a role in neutrophil migration and in the degranulation of neutrophils. Alternate translation initiation site usage, including a non-AUG (CUG) codon, results in the production of two different isoforms, that have different subcellular localization.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08631
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.02% NP-40, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 3055
Molecular Weight (g/mol) 84.8 kDa
Name Human HCK, GST Tag
Purification Method Purified
Quantity 10 μg
Gene Alias AI849071; B cell/myeloid kinase; B-cell/myeloid kinase; BMK; HCK; HCK proto-oncogene, Src family tyrosine kinase; Hctk; Hematopoietic cell kinase; hemopoietic cell kinase; hemopoietic cell tyrosine kinase; JTK9; p56Hck; p56-HCK; p56-HCK/p59-HCK; p59Hck; p59-HCK/p60-HCK; p61Hck; tyrosine-protein kinase HCK
Common Name HCK
Kinase Family SRC Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Protein Family Kinases & Inhibitors
Form Liquid
Protein Subtype Cytoplasmic Tyrosine Kinases
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.