Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HCK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA525989
Description
This antibody is predicted to react with mouse, non-human primate and rat based on sequence homology.
Hematopoietic cell kinase (HCK) is a SRC-related kinase expressed in lymphoid and hematopoietic cells and may play a role in some forms of leukemia. (HCK Alias: DKFZp686D0638). HCK is a protein-tyrosine kinase that is predominantly expressed in hemopoietic cell types. The encoded protein may help couple the Fc receptor to the activation of the respiratory burst. In addition, it may play a role in neutrophil migration and in the degranulation of neutrophils. Alternate translation initiation site usage, including a non-AUG (CUG) codon, results in the production of two different isoforms, that have different subcellular localization.
Specifications
HCK | |
Polyclonal | |
Unconjugated | |
HCK | |
AI849071; B cell/myeloid kinase; B-cell/myeloid kinase; BMK; HCK; HCK proto-oncogene, Src family tyrosine kinase; Hctk; Hematopoietic cell kinase; hemopoietic cell kinase; hemopoietic cell tyrosine kinase; JTK9; p56Hck; p56-HCK; p56-HCK/p59-HCK; p59Hck; p59-HCK/p60-HCK; p61Hck; tyrosine-protein kinase HCK | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
3055 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P08631 | |
HCK | |
KLH conjugated synthetic peptide between 131-156 amino acids from the N-terminal region of human HCK. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction