Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCLS1 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204028R
Description
HCLS1 Polyclonal antibody specifically detects HCLS1 in Human samples. It is validated for Western BlotSpecifications
HCLS1 | |
Polyclonal | |
Western Blot | |
CTTNL, hematopoietic cell-specific Lyn substrate 1hematopoietic lineage cell-specific protein, HS1lckBP1, LckBP1, p75 | |
The immunogen recognized by this antibody maps to a region between residue 350 and 400 of human Hematopoietic Cell-specific Lyn Substrate 1 using the numbering given in entry NP_005326.2 (GeneID 3059). | |
0.1 mL | |
Signal Transduction | |
3059 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction