Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HCLS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115346
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115346 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115346 Supplier Invitrogen™ Supplier No. PA5115346
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HCLS1.

Substrate of the antigen receptor-coupled tyrosine kinase. Plays a role in antigen receptor signaling for both clonal expansion and deletion in lymphoid cells. May also be involved in the regulation of gene expression. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCLS1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene HCLS1
Gene Accession No. P14317, P49710
Gene Alias AW213261; CTTNL; HCLS1; hematopoietic cell specific Lyn substrate 1; hematopoietic cell-specific Lyn substrate 1; Hematopoietic lineage cell-specific protein; HS1; LckBP1; p75
Gene Symbols HCLS1
Host Species Rabbit
Immunogen A synthesized peptide derived from human HCLS1(Accession P14317), corresponding to amino acid residues E371-G421.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15163, 288077, 3059
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.