Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HCLS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596079
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596079 Supplier Invitrogen™ Supplier No. PA596079
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MWKSVVGHDV SVSVETQGDD WDTDPDFVND ISEKEQRWGA KTIEGSGRTE HINIHQLRNK VSEEHDVLRK KEMESGPKAS HGYGGRFGVE RDRMDKSAVG HEYVAEVEKH SSQTDAAKGF GGKYGVERDR ADKSAVGFDY KGEVEKHTSQ KDYSRGFGGR YGVEKDKWDK AALGYDYKGE TEKHESQRDY AKGFGGQYGI QKDRVDKSAV GFNEMEAPTT AYKKTTPIEA ASSGTRGLKA KFESMAEEKR KREEEEKAQQ VARRQQERKA VTKRSPEAPQ; Positive Samples: BT-474, Jurkat; Cellular Location: Cytoplasm, Membrane, Mitochondrion, Peripheral membrane protein.

Substrate of the antigen receptor-coupled tyrosine kinase. Plays a role in antigen receptor signaling for both clonal expansion and deletion in lymphoid cells. May also be involved in the regulation of gene expression. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCLS1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HCLS1
Gene Accession No. P14317, P49710
Gene Alias AW213261; CTTNL; HCLS1; hematopoietic cell specific Lyn substrate 1; hematopoietic cell-specific Lyn substrate 1; Hematopoietic lineage cell-specific protein; HS1; LckBP1; p75
Gene Symbols HCLS1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human HCLS1 (NP_0053262).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15163, 3059
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.