Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HCMV IL-10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547082
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547082 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547082 Supplier Invitrogen™ Supplier No. PA547082
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, less than 1% cross-reactivity with recombinant mouse IL-10, recombinant canine IL-10, recombinant feline IL-10, recombinant cotton rat IL-10, recombinant rat IL-10, recombinant porcine IL-10, recombinant human IL-10, and recombinant Epstein-Barr virus encoded viral IL-10 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

Interleukin-10 (IL-10) is a pleiotropic cytokine that plays a crucial role in regulating immune and inflammstory responses. It has closely related homologs in the genome of several viruses including the human cytomegalovirus (HCMV), a prevalent pathogen that can remain in humans as a latent infection. Viruses use multiple strategies to avoid detection and clearance by the host immune system. One of these strategies is to express virus-encoded host cytokine homologs, such as IL-10, to modulate the host immune system to the virus' advantage. HCMV IL-10 is encoded from noncontiguous stretches in the viral genome and is transcribed early during infection. It shares 27% amino acid sequence identity with the endogenous human IL-10. Despite its low homology with the human protein, it can bind to the human IL-10 receptor complex and induces signal transduction. HCMV IL-10 has been shown to suppress cytokine production and inhibit proliferation of mitogen-stimulated human peripheral blood mononuclear cells. HCMV IL-10 can also down regulate the antigen-presenting and accessory cell functions of human monocytes. These immunosuppressive properties of HCMV IL-10 likely contribute to immune evasion during virus infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCMV IL-10
Applications Neutralization, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Alias CSIF; CSIFMGC126450; IL10A; interleukin 10; interleukin-10; TGIF
Host Species Goat
Immunogen E. coli-derived recombinant viral HCMV IL-10 Ala26-Lys176.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.