Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCN1 Antibody, Novus Biologicals™
SDP

Catalog No. NB018405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018405 0.025 mL
NBP120250 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018405 Supplier Novus Biologicals Supplier No. NBP1202500.025ML
Only null left
Add to Cart
Add to Cart

Sheep Polyclonal Antibody has been used in 6 publications

HCN1 Polyclonal specifically detects HCN1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HCN1
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence
Formulation Whole antisera with No Preservative
Gene Accession No. O60741
Gene Alias BCNG1, BCNG-1brain cyclic nucleotide gated channel 1, Brain cyclic nucleotide-gated channel 1, HAC-2, hyperpolarization activated cyclic nucleotide-gated potassium channel 1, potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1
Gene Symbols HCN1
Host Species Sheep
Immunogen A synthetic peptide from cytoplasmic region of human HCN1 conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous in many species including mouse and rat.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348980
Test Specificity HCN1.
Reconstitution Reconstitute with 0.1 ml sterilized water to desired concentration.
Target Species Mouse, Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.