Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCN1 Antibody (S70), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP122450
Description
HCN1 Monoclonal specifically detects HCN1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Microarray.Specifications
HCN1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1 - 10 ug/mL, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 0.1 - 1 ug/mL, Immunohistochemistry-Frozen 1:10 - 1:500, Microarray | |
HCN1 | |
Fusion protein amino acids 778-910 (C terminus) of rat HCN1 | |
0.1 mg | |
Primary | |
Detects approx. 100 kDa. No cross-reactivity against HCN2. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
S70 | |
Unconjugated | |
BCNG1, BCNG-1brain cyclic nucleotide gated channel 1, Brain cyclic nucleotide-gated channel 1, HAC-2, hyperpolarization activated cyclic nucleotide-gated potassium channel 1, potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1 | |
Mouse | |
Protein G purified | |
RUO | |
348980 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction