Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HCN1 Monoclonal Antibody (S70), APC
GREENER_CHOICE

Catalog No. PIMA545399
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545399 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545399 Supplier Invitrogen™ Supplier No. MA545399
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45399 was sufficient for detection of HCN1 in 10 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 100kDa. No cross-reactivity against HCN2.

HCN1 includes Hyperpolarization-activated cation channels of the HCN gene family, which contribute to spontaneous rhythmic activity in both heart and brain. HCN1 is a member of a family of pacemaker channels activated by hyperpolarization and regulated by cyclic nucleotides. HCN1 and HCN2 play an important role for motor learning and neuronal integration by cerebellar Purkinje cells as well as, shaping autonomous activity of single neurons and the periodicity of network oscillations. The HCN1 expression is highly enriched in cerebral cortex, hippocampus, cerebellum, and facial motor nucleus. HCN2 is highly abundant in mamillary bodies, pontine nucleus, ventral cochlear nucleus, and nucleus of the trapezoid body. These variations in regional specificity of HCN channels could generate important differences in neuronal pacemaker activity across brain systems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCN1
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone S70
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4
Gene HCN1
Gene Accession No. O60741, O88704, Q9JKB0
Gene Alias Bcng1; BCNG-1; Brain cyclic nucleotide-gated channel 1; C630013B14Rik; EIEE24; Hac2; HAC-2; Hcn1; hyperpolarization activated cyclic nucleotide gated potassium channel 1; hyperpolarization-activated cation channel 2; hyperpolarization-activated cyclic nucleotide-gated potassium channel 1; hyperpolarization-activated, cyclic nucleotide-gated K+ 1; potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1
Gene Symbols HCN1
Host Species Mouse
Immunogen Fusion protein amino acids 778-910 (C terminus) of rat HCN1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15165, 348980, 84390
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.