Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HCN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595716
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595716 Supplier Invitrogen™ Supplier No. PA595716
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse brain, Rat brain; Cellular Location: Cell membrane, Multi-pass membrane protein Immunogen sequence: PSLPHEVSTL ISRPHPTVGE SLASIPQPVT AVPGTGLQAG GRSTVPQRVT LFRQMSSGAI PPNRGVPPAP PPPAAALPRE SSSVLNTDPD AEKPRFASNL.

HCN1 includes Hyperpolarization-activated cation channels of the HCN gene family, which contribute to spontaneous rhythmic activity in both heart and brain. HCN1 is a member of a family of pacemaker channels activated by hyperpolarization and regulated by cyclic nucleotides. HCN1 and HCN2 play an important role for motor learning and neuronal integration by cerebellar Purkinje cells as well as, shaping autonomous activity of single neurons and the periodicity of network oscillations. The HCN1 expression is highly enriched in cerebral cortex, hippocampus, cerebellum, and facial motor nucleus. HCN2 is highly abundant in mamillary bodies, pontine nucleus, ventral cochlear nucleus, and nucleus of the trapezoid body. These variations in regional specificity of HCN channels could generate important differences in neuronal pacemaker activity across brain systems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCN1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene HCN1
Gene Accession No. O60741, O88704, Q9JKB0
Gene Alias Bcng1; BCNG-1; Brain cyclic nucleotide-gated channel 1; C630013B14Rik; EIEE24; Hac2; HAC-2; Hcn1; hyperpolarization activated cyclic nucleotide gated potassium channel 1; hyperpolarization-activated cation channel 2; hyperpolarization-activated cyclic nucleotide-gated potassium channel 1; hyperpolarization-activated, cyclic nucleotide-gated K+ 1; potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1
Gene Symbols HCN1
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15165, 348980, 84390
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.