Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCN2 Antibody (S71-37), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212895JF646
Description
HCN2 Monoclonal antibody specifically detects HCN2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)Specifications
HCN2 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
S71-37 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
BCNG2, BCNG-2, brain cyclic nucleotide gated channel 2, brain cyclic nucleotide-gated channel 2, HAC-1, hyperpolarization activated cyclic nucleotide-gated potassium channel 2, potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 2 | |
Fusion protein amino acids 761-863 of rat HCN2 | |
0.1 mL | |
Neuroscience | |
610 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction