Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HCN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142593
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142593 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142593 Supplier Invitrogen™ Supplier No. PA5142593
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

HCN3 is a member of the family of hyperpolarization-activated and cyclic-nucleotide-gated (HCN) channels. HCN currents have been linked to pacemaker activity in the heart and brain, resting potential control, as well as neuronal plasticity. HCN3 has been detected in the brain, but with less expression than other HCN channels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCN3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Hcn3
Gene Accession No. Q9JKA8
Gene Alias Bcng4; BCNG-4; Hac3; HAC-3; HCN3; Hcn4; hyperpolarization activated cyclic nucleotide gated potassium channel 3; hyperpolarization activated cyclic nucleotide-gated potassium channel 3; hyperpolarization-activated cation channel 3; hyperpolarization-activated cyclic nucleotide-gated potassium channel 3; hyperpolarization-activated, cyclic nucleotide-gated K+ 3; hyperpolarization-activated, cyclic nucleotide-gated K+ 4; KIAA1535; MGC131493; Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 3
Gene Symbols Hcn3
Host Species Goat
Immunogen Peptide with sequence C-RGVRGRAPSTGAQ.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114245
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.