Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCN3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989400
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9400 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9400 Supplier Bioss Supplier No. BS9856R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HCN3 is a member of the family of hyperpolarization-activated and cyclic-nucleotide-gated (HCN) channels. HCN currents have been linked to pacemaker activity in the heart and brain, resting potential control, as well as neuronal plasticity. HCN3 has been detected in the brain, but with less expression than other HCN channels.

Specifications

Antigen HCN3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Hcn3
Gene Accession No. Q9P1Z3
Gene Alias Bcng4; BCNG-4; Hac3; HAC-3; HCN3; Hcn4; hyperpolarization activated cyclic nucleotide gated potassium channel 3; hyperpolarization activated cyclic nucleotide-gated potassium channel 3; Hyperpolarization-activated cation channel 3; hyperpolarization-activated cyclic nucleotide-gated potassium channel 3; hyperpolarization-activated, cyclic nucleotide-gated K+ 3; hyperpolarization-activated, cyclic nucleotide-gated K+ 4; KIAA1535; MGC131493; Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 3
Gene Symbols Hcn3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HCN3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57657
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.