Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCN4 Antibody (S114/10), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212910MFV450
Description
HCN4 Monoclonal antibody specifically detects HCN4 in Human, Mouse, Rat, Rabbit samples. It is validated for Flow CytometrySpecifications
HCN4 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Rabbit | |
Purified |
Flow Cytometry | |
S114/10 | |
Flow Cytometry | |
hyperpolarization activated cyclic nucleotide-gated cation channel 4, hyperpolarization activated cyclic nucleotide-gated potassium channel 4, potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4, SSS2 | |
Fusion protein amino acids 1019-1198 (c-terminus) of rat HCN4 | |
0.1 mL | |
Neuroscience | |
10021 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction