Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCN4 Antibody (SHG 1E5), Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10074439
Description
HCN4 Monoclonal specifically detects HCN4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence.Specifications
HCN4 | |
Monoclonal | |
Unconjugated | |
Q9Y3Q4 | |
HCN4 | |
Synthetic peptide sequence corresponding to residues S H G S L L L P P A S S P P P P Q V P Q R R G T P P L T P G R L T Q D L K L of HCN4. | |
100 μL | |
Primary | |
HCN4 (SHG 1E5) | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
SHG 1E5 | |
Western Blot 1:5000, Flow Cytometry 1:50, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:100, Immunohistochemistry-Frozen 1:1000, Immunofluorescence 1:10 - 1:500 | |
hyperpolarization activated cyclic nucleotide-gated cation channel 4, hyperpolarization activated cyclic nucleotide-gated potassium channel 4, potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4, SSS2 | |
Rat | |
Unpurified | |
RUO | |
10021 | |
Human, Mouse, Rat | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction