Learn More
Invitrogen™ HCN4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5143877
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, mouse brain tissue. Flow: MCF-7 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
HCN4 is a member of the family of hyperpolarization-activated and cyclic-nucleotide-gated (HCN) channels. HCN currents have been linked to pacemaker activity in the heart and brain, resting potential control, as well as neuronal plasticity. It has been shown that HCN4 channels function as receptors for sour taste, and are associated with pacemaker potential generation in the sinoatrial node.
Specifications
HCN4 | |
Polyclonal | |
Unconjugated | |
Hcn4 | |
Bcng3; BCNG-3; brain cyclic nucleotide-gated channel 3; Hcn3; Hcn4; hyperpolarization activated cyclic nucleotide gated potassium channel 4; hyperpolarization activated cyclic nucleotide-gated cation channel 4; hyperpolarization activated cyclic nucleotide-gated potassium channel 4; hyperpolarization-activated, cyclic nucleotide-gated K+ 3; hyperpolarization-activated, cyclic nucleotide-gated K+ 4; potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4; SSS2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10021, 330953, 59266 | |
-20°C | |
Lyophilized |
ELISA, Flow Cytometry, Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and no preservative | |
O70507, Q9JKA7, Q9Y3Q4 | |
Hcn4 | |
E.coli-derived human HCN4 recombinant protein (Position: H909-L1203). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.